Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

674125

Sigma-Aldrich

Salicylaldehyde thiosemicarbazone palladium(II) chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8ClN3OPdS
CAS Number:
Molecular Weight:
336.11
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

>300 °C

SMILES string

NC(N)=S.Cl[Pd]1Oc2ccccc2C=N1

InChI

1S/C7H6NO.CH4N2S.ClH.Pd/c8-5-6-3-1-2-4-7(6)9;2-1(3)4;;/h1-5,9H;(H4,2,3,4);1H;/q-1;;;+3/p-2

InChI key

PHBJCXMVMUSQHT-UHFFFAOYSA-L

Application

Catalyst used for Suzuki Coupling under aerobic conditions[1]

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kostas, I. D. et al.
Tetrahedron Letters, 46, 1967-1967 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service