Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

674052

Sigma-Aldrich

LaPCat Perovskite Catalyst Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300

reaction suitability

reagent type: catalyst
core: palladium

Quality Level

Application

A kit of 6 palladium and/or copper heterogeneous perovskite catalysts which facilitate aryl-aryl cross coupling reactions.

Legal Information

LaPCat is a trademark of Reaxa Ltd.

Kit Components Only

Product No.
Description

  • LaPCat FP2 100 mg

  • LaPCat FP8 100 mg

  • LaPCat CoP 100 mg

  • LaPCat CuP 100 mg

  • LaPCat CoCu 100 mg

  • LaPCat YBCu 100 mg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service