Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

673900

Sigma-Aldrich

Ethyl N-Boc-4-(4-chlorobenzyl)piperidine-4-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28ClNO4
CAS Number:
Molecular Weight:
381.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

69-73 °C

functional group

chloro
ester

SMILES string

CCOC(=O)C1(CCN(CC1)C(=O)OC(C)(C)C)Cc2ccc(Cl)cc2

InChI

1S/C20H28ClNO4/c1-5-25-17(23)20(14-15-6-8-16(21)9-7-15)10-12-22(13-11-20)18(24)26-19(2,3)4/h6-9H,5,10-14H2,1-4H3

InChI key

QTVIWIWJGWSTDC-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service