About This Item
assay
97%
refractive index
n20/D 1.534
density
1.9802 g/mL at 25 °C
SMILES string
FC(F)(F)c1ccc(I)c(Cl)c1
InChI
1S/C7H3ClF3I/c8-5-3-4(7(9,10)11)1-2-6(5)12/h1-3H
InChI key
ULJRBVGSKAKZKQ-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
>230.0 °F - closed cup
flash_point_c
> 110 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service