673633
Decanethiol functionalized silver nanoparticles
3-7 nm particle size (TEM), 0.1 % (w/v) in hexane
Synonym(s):
Decylthiol functionalized silver nanoparticles
About This Item
Recommended Products
form
nanoparticles
solution
concentration
0.1 % (w/v) in hexane
extent of labeling
0.075-0.125% loading
particle size
3-7 nm (TEM)
density
0.672 g/mL at 25 °C
functional group
thiol
signalword
Danger
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3
target_organs
Central nervous system, Nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-14.8 °F - closed cup
flash_point_c
-26 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service