Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

672769

Sigma-Aldrich

Methyl phthaloyl chloride

97%

Synonym(s):

2-Chlorocarbonylbenzoic acid methyl ester, Methyl 2-(chlorocarbonyl)benzoate, Methyl 2-(chloroformyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClO3
CAS Number:
Molecular Weight:
198.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥96.5% (HPLC)
96.5-103.5% (AT)
97%

form

liquid

functional group

acyl chloride
ester

storage temp.

2-8°C

SMILES string

COC(=O)c1ccccc1C(Cl)=O

InChI

1S/C9H7ClO3/c1-13-9(12)7-5-3-2-4-6(7)8(10)11/h2-5H,1H3

InChI key

GNBWUEAMCSHHMO-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service