672602
3,7-Dioxa-1,9-nonanediol
≥98.0% (GC)
Synonym(s):
1,9-Dihydroxy-3,7-dioxanonane, 2,2′-(Trimethylenedioxy)diethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H16O4
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
≥98.0% (GC)
refractive index
n20/D 1.456
density
1.091 g/mL at 20 °C
functional group
ether
hydroxyl
SMILES string
OCCOCCCOCCO
InChI
1S/C7H16O4/c8-2-6-10-4-1-5-11-7-3-9/h8-9H,1-7H2
InChI key
KXSKAZFMTGADIV-UHFFFAOYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service