Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672408

Sigma-Aldrich

(R)-N,N′-Dimethyl-1,1′-binaphthyldiamine

≥99.0%

Synonym(s):

(R)-N,N′-Dimethyl-2,2′-diamino-1,1′-binaphthyl, (R)-N,N′-Dimethyl-1,1′-binaphthalene-2,2′-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2
CAS Number:
Molecular Weight:
312.41
Beilstein/REAXYS Number:
5588762
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥99.0% (HPLC)
≥99.0%

optical purity

enantiomeric excess: ≥99.0% (HPLC)

storage temp.

2-8°C

SMILES string

CNc1ccc2ccccc2c1-c3c(NC)ccc4ccccc34

InChI

1S/C22H20N2/c1-23-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)24-2/h3-14,23-24H,1-2H3

InChI key

PXDKDUXMEHPNCO-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service