Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

671428

Sigma-Aldrich

(S)-α,α-Bis(3,5-dimethylphenyl)-2-pyrrolidinemethanol trimethylsilyl ether hydrochloride

97%

Synonym(s):

(S)-2-[Bis(3,5-dimethylphenyl)(trimethylsilyloxy)methyl]pyrrolidine hydrochloride, (S)a,a-Bis(3,5-dimethylphenyl)prolinol trimethylsilyl ether hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H35NOSi · HCl
Molecular Weight:
418.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥96.5% (HPLC)
97%

optical purity

enantiomeric excess: ≥96.5% (HPLC)

mp

159-162 °C

storage temp.

2-8°C

SMILES string

Cl.Cc1cc(C)cc(c1)C(O[Si](C)(C)C)([C@@H]2CCCN2)c3cc(C)cc(C)c3

InChI

1S/C24H35NOSi.ClH/c1-17-11-18(2)14-21(13-17)24(26-27(5,6)7,23-9-8-10-25-23)22-15-19(3)12-20(4)16-22;/h11-16,23,25H,8-10H2,1-7H3;1H/t23-;/m0./s1

InChI key

WTDMRDKYLPARGO-BQAIUKQQSA-N

Application

Catalyst for enantioselective ene reaction[1]; dienamine catalysis[2]; catalyst for alpha-sulfenylation of aldehydes[3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective ene reaction of cyclopentadiene and alpha,beta-enals catalyzed by a diphenylprolinol silyl ether.
Hiroaki Gotoh et al.
Angewandte Chemie (International ed. in English), 45(41), 6853-6856 (2006-09-27)
Søren Bertelsen et al.
Journal of the American Chemical Society, 128(39), 12973-12980 (2006-09-28)
A new concept in organocatalysis is presented, the direct asymmetric gamma-functionalization of alpha,beta-unsaturated aldehydes. We disclose that secondary amines can invert the usual reactivity of alpha,beta-unsaturated aldehydes, enabling a direct gamma-amination of the carbonyl compound using azodicarboxylates as the electrophilic
Enantioselective organocatalyzed alpha sulfenylation of aldehydes.
Mauro Marigo et al.
Angewandte Chemie (International ed. in English), 44(5), 794-797 (2005-01-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service