Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

669180

Sigma-Aldrich

(R)-(−)-3,3′-Bis(3,5-dimethylphenyl)-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-2-naphthol

97%

Synonym(s):

(R)-3,3′-Bis(3,5-dimethylphenyl)-5,6,7,8,5′,6′,7′,8′-octahydro-[1,1′]binaphthalenyl-2, 2′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H38O2
Molecular Weight:
502.69
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

218-222 °C

InChI

1S/C36H38O2/c1-21-13-22(2)16-27(15-21)31-19-25-9-5-7-11-29(25)33(35(31)37)34-30-12-8-6-10-26(30)20-32(36(34)38)28-17-23(3)14-24(4)18-28/h13-20,37-38H,5-12H2,1-4H3

InChI key

RSHMDLJRFPHHRE-UHFFFAOYSA-N

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

TADDOLs demonstrate versatility in metal-catalyzed asymmetric reactions and as Brønsted acid organocatalysts in hetero-Diels–Alder reactions.

We present an article concerning BINOL and Derivatives.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service