Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
668885
Bis(tert-butylimino)bis(dimethylamino)tungsten(VI)
packaged for use in deposition systems
Synonym(s):
BTBMW
About This Item
Recommended Products
assay
99.99% (trace metals analysis)
form
liquid
reaction suitability
core: tungsten
bp
81 °C/0.02 mmHg (lit.)
density
1.305 g/mL at 25 °C (lit.)
SMILES string
CN(C)[W](=NC(C)(C)C)(=NC(C)(C)C)N(C)C
InChI
1S/2C4H9N.2C2H6N.W/c2*1-4(2,3)5;2*1-3-2;/h2*1-3H3;2*1-2H3;/q;;2*-1;+2
InChI key
JVCWKXBYGCJHDF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Water-react 1
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
77.9 °F - closed cup - (own results)
flash_point_c
25.5 °C - closed cup - (own results)
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.
High Purity Metalorganic Precursors for CPV Device Fabrication
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service