Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668605

Sigma-Aldrich

QuadraPure® BDZ

400-750 μm, extent of labeling: 1.0-1.4 mmol/g loading, macroporous

Synonym(s):

QuadraPure® Bis(propyl-1H-imidazole)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

macroporous

reaction suitability

reagent type: catalyst
reagent type: chelator

extent of labeling

1.0-1.4 mmol/g loading

particle size

400-750 μm

Application

Metal Scavenger: Rh, Co, Pd, Ni

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service