Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668583

Sigma-Aldrich

QuadraPure® EDA

500-800 μm, extent of labeling: 1.5-2.0 mmol/g loading, macroporous

Synonym(s):

QuadraPure® Bis(ethylamine)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
PubChem Substance ID:

product line

QuadraPure®

form

macroporous

reaction suitability

reaction type: solution phase peptide synthesis
reagent type: catalyst
reagent type: chelator

extent of labeling

1.5-2.0 mmol/g loading

particle size

500-800 μm

functional group

amine

Application

Metal Scaventer: Pd in basic media and with phosphines, Co, Ni, Rh

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dean Brady et al.
BMC biotechnology, 8, 8-8 (2008-02-02)
Enzymes have found extensive and growing application in the field of chemical organic synthesis and resolution of chiral intermediates. In order to stabilise the enzymes and to facilitate their recovery and recycle, they are frequently immobilised. However, immobilisation onto solid
QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service