Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667951

Sigma-Aldrich

(4-Methoxyphenyl)dimethylsilanol

96%

Synonym(s):

(p-Methoxyphenyl)dimethylsilanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3O)C6H4)Si(CH3)2OH
CAS Number:
Molecular Weight:
182.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.522

density

1.101 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)[Si](C)(C)O

InChI

1S/C9H14O2Si/c1-11-8-4-6-9(7-5-8)12(2,3)10/h4-7,10H,1-3H3

InChI key

DDLYKNBMTGBOQY-UHFFFAOYSA-N

Application

(4-Methoxyphenyl)dimethylsilanol can be used:
  • As a starting material in the Pd-catalyzed cross-coupling reactions with various aryl halides to obtain biaryl compounds using Cs2CO3 base.[1][2][3]
  • As a starting material to prepare potassium (4-methoxyphenyl)dimethylsilanolate, which in turn is used to synthesize biaryl derivatives by reacting with aryl bromides.[4]
  • As a co-catalyst along with Mo(CO)6 applicable in the 4-decyne metathesis.[5]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Cross-Coupling Reactions of Substituted Aryl (dimethyl) silanols
Denmark SE and Ober MH
advanced synthesis and catalysis, 346(13-15), 1703-1714 (2004)
Phosphine oxides as stabilizing ligands for the palladium-catalyzed cross-coupling of potassium aryldimethylsilanolates
Denmark SE, et al.
Tetrahedron, 63(26), 5730-5738 (2007)
Organosilanes for Cross-coupling
Aldrich Chemfiles, 6(5), 7-9 (2006)
Cross-coupling reactions of arylsilanols with substituted aryl halides
Denmark SE and Ober MH
Organic Letters, 5(8), 1357-1360 (2003)
Palladium-Catalyzed Cross-Coupling Reactions of Substituted Aryl (dimethyl) silanols
Denmark SE and Ober MH
Advanced Synthesis & Catalysis, 346(13-15), 1703-1714 (2004)

Articles

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service