Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667587

Sigma-Aldrich

(7R,10S)-(+)-1-Aza-10-isopropyl-8-oxa-4-thiabicyclo[5.3.0]-2-decanone

97%

Synonym(s):

Gleason chiral auxiliary

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO2S
Molecular Weight:
215.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

142-146 °C

SMILES string

[H][C@@]12CCSCC(=O)N1[C@H](CO2)C(C)C

InChI

1S/C10H17NO2S/c1-7(2)8-5-13-10-3-4-14-6-9(12)11(8)10/h7-8,10H,3-6H2,1-2H3/t8-,10-/m1/s1

InChI key

DCIDQFQEFRCDOM-PSASIEDQSA-N

Application

This chiral lactam auxiliary has been successfully employed in the stereocontrolled synthesis of all carbon quaternary centers. The unique methodology developed by Gleason allows for the creation of either antipode of the product using a single isomer of the auxiliary.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azélie Arpin et al.
Organic letters, 8(7), 1359-1362 (2006-03-28)
[reaction: see text] Chiral bicyclic thioglycolate lactams may be prepared in three steps from inexpensive commercial materials. The resulting lactams may be alkylated three times, twice using basic enolization and once using reductive enolization, to form alpha-quaternary carboxylic acid derivatives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service