Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667471

Sigma-Aldrich

Anisotropic Silicon <100> Etchant

Synonym(s):

Preferential Silicon Etchant, Silicon 100 Etch, Silicon Preferential Etch

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.23

refractive index

n20/D 1.484

bp

>240 °C

density

1.001 g/mL at 25 °C

General description

Ethylenediamine based - for < 100 > crystal orientation

Application

Etch rate ~80microns/hour at @ 100 °C. Etches cleanly with only a deionized water rinse needed. Will not etch Au or Ta.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

109.9 °F

flash_point_c

43.3 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service