Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667463

Sigma-Aldrich

Titanium etchant

Synonym(s):

Titanium etch

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300

vapor pressure

14 mmHg ( 20 °C)

bp

200 °C

mp

−35 °C

Application

Designed for etching evaporated films commonly employed as bonding and barrier layers in microelectronics. Excellent resolution, photo resist compatibility, and minimal undercutting are readily achieved.

Features and Benefits

Use with Polyethylene tanks between 20°-50°C.
Etch Rates
25Å/sec @ 20°C,
50Å/sec @ 30°C.

Rinse with DI water

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service