Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667455

Sigma-Aldrich

Anisotropic Silicon <110> Etchant

Synonym(s):

Preferential Silicon Etchant, Silicon 110 Etch, Silicon Preferential Etch

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
Product 667455 is not currently sold in your country. Contact Technical Service

General description

Alkali based - for < 110 > crystal orientation - etches with vertical walls.

Application

Etch rate 1 mil/3 min at @ 100 °C. Etches cleanly with only a deionized water rinse needed. Will not etch Au or SiO2

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service