Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667161

Sigma-Aldrich

Potassium (3,3-dimethylbutyl)trifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH2BF3K
Molecular Weight:
192.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

95%

form

solid

mp

>300 °C

SMILES string

[K+].CC(C)(C)CC[B-](F)(F)F

InChI

1S/C6H13BF3.K/c1-6(2,3)4-5-7(8,9)10;/h4-5H2,1-3H3;/q-1;+1

InChI key

RAXISKMTAMCZKQ-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service