Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667099

Sigma-Aldrich

Dimethyl(2-thienyl)silanol

97%

Synonym(s):

Dimethyl(thien-2-yl)silanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H3S)Si(OH)(CH3)2
CAS Number:
Molecular Weight:
158.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.521

density

1.085 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(O)c1cccs1

InChI

1S/C6H10OSSi/c1-9(2,7)6-4-3-5-8-6/h3-5,7H,1-2H3

InChI key

UDIQRZJWIBRPFX-UHFFFAOYSA-N

Application

A useful silicon nucleophile for Pd-catalyzed cross-coupling.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service