Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

666645

Sigma-Aldrich

9-(Tetrahydro-2H-pyran-2-yloxy)-7-nonynoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O4
CAS Number:
Molecular Weight:
254.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.484

density

1.081 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCC#CCOC1CCCCO1

InChI

1S/C14H22O4/c15-13(16)9-5-3-1-2-4-7-11-17-14-10-6-8-12-18-14/h14H,1-3,5-6,8-12H2,(H,15,16)

InChI key

BNWVOWGHORIPKA-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service