Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665983

Sigma-Aldrich

5-Chloro-2-thiophenecarbonitrile

97%

Synonym(s):

2-Chloro-5-cyanothiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2ClNS
CAS Number:
Molecular Weight:
143.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.587

bp

208 °C/760 mmHg

density

1.361 g/mL at 25 °C

SMILES string

Clc1ccc(s1)C#N

InChI

1S/C5H2ClNS/c6-5-2-1-4(3-7)8-5/h1-2H

InChI key

MZALEEOEVCTDJA-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

204.1 °F

flash_point_c

95.6 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service