Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665754

Sigma-Aldrich

4-Pentylphenyl 4-methylbenzoate

97%

Synonym(s):

p-Amylphenyl p-toluate, p-Pentylphenyl p-methylbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid crystal

mp

34-38 °C

storage temp.

2-8°C

SMILES string

CCCCCc1ccc(OC(=O)c2ccc(C)cc2)cc1

InChI

1S/C19H22O2/c1-3-4-5-6-16-9-13-18(14-10-16)21-19(20)17-11-7-15(2)8-12-17/h7-14H,3-6H2,1-2H3

InChI key

OFNFLSYTTLXGBM-UHFFFAOYSA-N

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service