Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665134

Sigma-Aldrich

Oxotrichloro[bis(diphenylphosphino)methane]rhenium(V)

Synonym(s):

Trichloro[methylenebis[diphenylphosphine]-P,P′]oxorhenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22Cl3OP2Re
CAS Number:
Molecular Weight:
692.96
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: rhenium
reagent type: catalyst

SMILES string

Cl[Re](Cl)(Cl)=O.C(P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C25H22P2.3ClH.O.Re/c1-5-13-22(14-6-1)26(23-15-7-2-8-16-23)21-27(24-17-9-3-10-18-24)25-19-11-4-12-20-25;;;;;/h1-20H,21H2;3*1H;;/q;;;;;+3/p-3

InChI key

SAVOOVLDSRTAGJ-UHFFFAOYSA-K

Application

Catalyst for:
  • Epoxidation of olefins using hydrogen peroxide activated bicarbonate as an oxidant
  • Efficient reduction of aromatic sulfoxides
  • Regioselective coupling of propargyl alcohols with sulfonamides and carbamates
  • Cycloisomerization reactions

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service