Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

664979

Sigma-Aldrich

Dichlorobis(2-(diphenylphosphino)ethylamine)ruthenium(II)

95%

Synonym(s):

Dichlorobis[2-(diphenylphosphino-κP)ethanamine-κN]ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32Cl2N2P2Ru
CAS Number:
Molecular Weight:
630.49
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

130-135 °C

SMILES string

Cl[Ru]Cl.NCCP(c1ccccc1)c2ccccc2.NCCP(c3ccccc3)c4ccccc4

InChI

1S/2C14H16NP.2ClH.Ru/c2*15-11-12-16(13-7-3-1-4-8-13)14-9-5-2-6-10-14;;;/h2*1-10H,11-12,15H2;2*1H;/q;;;;+2/p-2

InChI key

BJNVYFXBTNBKRU-UHFFFAOYSA-L

Application

Catalyst for:
  • Synthesis of long chain linear fatty acid esters
  • Preparation of ruthenium borohydride complexes for use in hydrogenation reactions
  • Dehydrogenation of ammonia boranes
  • Chemoselective reduction of carboxylic esters
  • Synthesis of ruthenium beta-aminophosphine hydrido/chloro complexes with catalytic activities

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Aminophosphine Ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service