Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

664871

Sigma-Aldrich

trans-2-(2,4-Difluorophenyl)vinylboronic acid pinacol ester

96%

Synonym(s):

2-(E-2-(2,4-Difluorophenyl)vinyl)-4,4,5,5-tetramethyl[1,3,2]dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17BF2O2
CAS Number:
Molecular Weight:
266.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.503

bp

103-105 °C/0.2-0.3 mmHg

density

1.099 g/mL at 25 °C

SMILES string

CC1(C)OB(OC1(C)C)\C=C\c2ccc(F)cc2F

InChI

1S/C14H17BF2O2/c1-13(2)14(3,4)19-15(18-13)8-7-10-5-6-11(16)9-12(10)17/h5-9H,1-4H3/b8-7+

InChI key

BMZXZTQQERUFEM-BQYQJAHWSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service