Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

663298

Sigma-Aldrich

4-Methylbenzylboronic acid pinacol ester

97%

Synonym(s):

2-(4-Methylbenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21BO2
CAS Number:
Molecular Weight:
232.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.489

bp

95 °C/1 mmHg

density

0.953 g/mL at 25 °C

SMILES string

Cc1ccc(CB2OC(C)(C)C(C)(C)O2)cc1

InChI

1S/C14H21BO2/c1-11-6-8-12(9-7-11)10-15-16-13(2,3)14(4,5)17-15/h6-9H,10H2,1-5H3

InChI key

IRKUSKILEFQOJQ-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service