Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

663247

Sigma-Aldrich

1,3,5-Tris(diphenylamino)benzene

greener alternative

97%

Synonym(s):

TDAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H33N3
CAS Number:
Molecular Weight:
579.73
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

252-256 °C

greener alternative category

SMILES string

c1ccc(cc1)N(c2ccccc2)c3cc(cc(c3)N(c4ccccc4)c5ccccc5)N(c6ccccc6)c7ccccc7

InChI

1S/C42H33N3/c1-7-19-34(20-8-1)43(35-21-9-2-10-22-35)40-31-41(44(36-23-11-3-12-24-36)37-25-13-4-14-26-37)33-42(32-40)45(38-27-15-5-16-28-38)39-29-17-6-18-30-39/h1-33H

InChI key

DPFGGYIWNDCEJM-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". 1,3,5-Tris(diphenylamino)benzeneis a hole transporting material and an electron-blocking layer, has high hole mobility, low electron affinity, and high ionic potential. Its electron blocking nature results in effective confinement of injected charge carriers in the perovskite layers. Click here for more information.

application

Hole transport material employed in electroluminescent devices.[1] The radical cation is used in molecular magnetics.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yoshizawa, K.;
Bulletin of the Chemical Society of Japan, 69, 1417-1422 (1996)
Ishikawa, W.;
Chemistry Letters (Jpn), 10, 173-174 (1991)
Miao Song et al.
Science (New York, N.Y.), 367(6473), 40-45 (2019-11-30)
Natural and synthetic nanoparticles composed of fivefold twinned crystal domains have distinct properties. The formation mechanism of these fivefold twinned nanoparticles is poorly understood. We used in situ high-resolution transmission electron microscopy combined with molecular dynamics simulations to demonstrate that
Duangkamol Lewchalermwong et al.
Animal reproduction science, 223, 106622-106622 (2020-10-20)
The gonadotropin compound, PG600, is used to induce estrus in prepubertal gilts, but responses can be variable. This study was conducted to evaluate PG600 effects on follicles, estrus, ovulation and estrogen production. Prepubertal gilts (n = 50) were treated with PG600. Gilts
Jaewon Lee et al.
ACS nano, 12(12), 12778-12787 (2018-11-14)
Superlattice structures formed by nanoparticle (NP) self-assembly have attracted increasing attention due to their potential as a class of nanomaterials with enhanced physicochemical properties tailored by the assembly structure. However, many key questions remain regarding the correlation between the dynamics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service