Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662992

Sigma-Aldrich

trans-1-Hepten-1-ylboronic acid pinacol ester

97%

Synonym(s):

E-2-(1-Heptenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-1-Heptenylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H25BO2
CAS Number:
Molecular Weight:
224.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.445

bp

73-77 °C/0.4-0.5 mmHg

density

0.875 g/mL at 25 °C

SMILES string

CCCCC\C=C\B1OC(C)(C)C(C)(C)O1

InChI

1S/C13H25BO2/c1-6-7-8-9-10-11-14-15-12(2,3)13(4,5)16-14/h10-11H,6-9H2,1-5H3/b11-10+

InChI key

XHEDFAYNMNXKGM-ZHACJKMWSA-N

Application

Substrate used in a palladium-catalyzed cross-coupling with olefins providing substituted 1,3-dienes.[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Soo Yoo et al.
Journal of the American Chemical Society, 128(50), 16384-16393 (2006-12-15)
We report herein the development of a general and mild protocol of oxygen-promoted Pd(II) catalysis resulting in the selective cross-couplings of alkenyl- and arylboron compounds with various olefins. Unlike most cross-coupling reactions, this new methodology works well even in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service