Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662917

Sigma-Aldrich

1,7-Heptanediol-1,3,5,7-13C4

95% (CP), 99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C3H16O2
CAS Number:
Molecular Weight:
136.17
UNSPSC Code:
12352001

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

assay

95% (CP)

mass shift

M+3

SMILES string

O[13CH2]C[13CH2]C[13CH2]C[13CH2]O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service