Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

662593

Sigma-Aldrich

Lumidot CdS-6, quantum dot nanoparticles kit

core-type, fluorescence λem 380-480 nm

Synonym(s):

QD, Quantum dot semiconductor nanocrystal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

form

solution

shelf life

1 yr

concentration

5 mg/mL in toluene

bp

110 °C

density

0.865 g/mL at 25 °C

fluorescence

FWHM <30 nm
λem 380-480 nm

storage temp.

2-8°C

General description

Organically stabilized cadmium sulfide quantum dots in toluene.
Kit contains six toluene solutions of CdS quantum dot nanocrystals with fluorescence emission maxima spanning UV-blue spectrum:
  • Lumidot CdS 380 5 ml, 5 mg/ml, 360 nm
  • Lumidot CdS 400 5 ml, 5 mg/ml, 380 nm
  • Lumidot CdS 420 5 ml, 5 mg/ml, 400 nm
  • Lumidot CdS 440 5 ml, 5 mg/ml, 420 nm
  • Lumidot CdS 460 5 ml, 5 mg/ml, 440 nm
  • Lumidot CdS 480 5 ml, 5 mg/ml, 460 nm
Lumidot CdS 380-480 nanocrystals are surface-stabilized with oleic acid coating.
Typical nanocrystal sizes range from 1.6 nm (Lumidot CdS 380) to 7.3 nm (Lumidot CdS 480). The soft ligands increase hydrodynamic diameter of the nanocrystals by ~4.5-5 nm.

Features and Benefits

The coating has a surface stabilized with oleic acid ligands.

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents and patents pending.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Lumidot CdS, core-type quantum dots, 5 mg/mL in toluene 5 mL

  • Lumidot CdS, 440, core-type quantum dots, 5 mg/mL in toluene 5 mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • 662429Lumidot CdS 380 5 mLSDS

  • 662429Lumidot CdS 400 5 mLSDS

  • Lumidot CdS 440 5 mL

  • Lumidot CdS 480 5 mL

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

39.2 °F

flash_point_c

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service