Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662453

Sigma-Aldrich

2-Methoxyphenylmagnesium bromide solution

1.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4MgBr
CAS Number:
Molecular Weight:
211.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.791 g/mL at 25 °C

SMILES string

COc1ccccc1[Mg]Br

InChI

1S/C7H7O.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h2-5H,1H3;1H;/q;;+1/p-1

InChI key

BJJFGIPCBFILPB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service