Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

660388

Sigma-Aldrich

CX11

Umicore

Synonym(s):

1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene(1,4-naphthoquinone)palladium(0) dimer, Naphthoquinone-1,3-bis(2,6-diisopropylphenyl)-imidazole-2-ylidene-palladium(0) dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H84N4O4Pd2
CAS Number:
Molecular Weight:
1306.32
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

>300 °C

SMILES string

O=C1C=CC(=O)c2ccccc12.O=C3C=CC(=O)c4ccccc34.CC(C)c5cccc(C(C)C)c5N6C=CN(C6[Pd])c7c(cccc7C(C)C)C(C)C.CC(C)c8cccc(C(C)C)c8N9C=CN(C9[Pd])c%10c(cccc%10C(C)C)C(C)C

InChI

1S/2C27H37N2.2C10H6O2.2Pd/c2*1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;2*11-9-5-6-10(12)8-4-2-1-3-7(8)9;;/h2*9-21H,1-8H3;2*1-6H;;

InChI key

SQNPUPJPLWQFHN-UHFFFAOYSA-N

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In collaboration with Umicore AG and Co.,1 we are pleased to offer a series of robust Pd(II) and Pd(0) complexes employed as the linchpin in C–C bond forming reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service