Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

660205

Sigma-Aldrich

N-Boc-(2S,3S)-(−)-2-amino-3-methyl-1-pentanol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NO3
CAS Number:
Molecular Weight:
217.31
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.455

bp

121-126 °C/0.6 mmHg

SMILES string

CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C11H23NO3/c1-6-8(2)9(7-13)12-10(14)15-11(3,4)5/h8-9,13H,6-7H2,1-5H3,(H,12,14)/t8-,9+/m0/s1

InChI key

BPLDQMXXYMKQPW-DTWKUNHWSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service