Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658774

Sigma-Aldrich

Salicylaldehyde thiosemicarbazone

95%

Synonym(s):

2-Hydroxybenzaldehyde thiosemicarbazone, 2-[(2-Hydroxyphenyl)methylene]hydrazinecarbothioamide, o-Hydroxybenzaldehyde thiosemicarbazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=NNHC(S)NH2
CAS Number:
Molecular Weight:
195.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

solid

mp

>230 °C

SMILES string

NC(=S)N\N=C/c1ccccc1O

InChI

1S/C8H9N3OS/c9-8(13)11-10-5-6-3-1-2-4-7(6)12/h1-5,12H,(H3,9,11,13)/b10-5+

InChI key

OHNSRYBTSVDDKA-BJMVGYQFSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pabla Noblía et al.
Journal of inorganic biochemistry, 99(2), 443-451 (2004-12-29)
As a contribution to the development of novel vanadium complexes with pharmacologically interesting moieties, new dioxovanadium(V) semicarbazone complexes with the formula cis-VO(2)L, where L=5-bromosalicylaldehyde semicarbazone and 2-hydroxynaphtalen-1-carboxaldehyde semicarbazone have been synthesized and characterized by (1)H and (13)C NMR, Raman and
Kevin J Duffy et al.
Journal of medicinal chemistry, 45(17), 3573-3575 (2002-08-09)
High-throughput screening has resulted in the discovery of thiosemicarbazone thrombopoietin mimics. A shared pharmacophore hypothesis between this series and a previously identified class, the pyrazol-4-ylidenehydrazines, led to the rapid optimization of both potency and efficacy of the thiosemicarbazones. The application
R Prabhakaran et al.
Metallomics : integrated biometal science, 3(1), 42-48 (2010-12-07)
Bis salicylaldehyde-4(N)-ethylthiosemicarbazone ruthenium(iii) triphenylphosphine [Ru(Sal-etsc)(H-Sal-etsc)(PPh(3))] was synthesized and structurally characterized by spectral and X-ray crystallographic studies and it showed 100% inhibition on the DPPH radical. It also exhibited a significant lymphocyte activity and inhibitory effect on the lung carcinoma A549
Eswaran Ramachandran et al.
Metallomics : integrated biometal science, 4(2), 218-227 (2011-11-22)
New complexes, [Ni(HL)(PPh(3))]Cl (1), [Pd(L)(PPh(3))](2), and [Pd(L)(AsPh(3))](3), were synthesized from the reactions of 4-chloro-5-methyl-salicylaldehyde thiosemicarbazone [H(2)L] with [NiCl(2)(PPh(3))(2)], [PdCl(2)(PPh(3))(2)] and [PdCl(2)(AsPh(3))(2)]. They were characterized by IR, electronic, (1)H-NMR spectral data. Further, the structures of the complexes have been determined by
D Chattopadhyay et al.
Acta crystallographica. Section C, Crystal structure communications, 44 ( Pt 6), 1025-1028 (1988-06-15)
C8H9N3OS, monoclinic, C2/c, a = 14.206 (3), b = 14.244 (4), c = 10.457 (4) A, beta = 116.18(2) degrees, V = 1898.9 (8) A3, Z = 8, Dm = 1.387, D chi = 1.366 g cm-3, lambda(Mo K alpha)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service