Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65820

Sigma-Aldrich

Methyl N-methylanthranilate

technical, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3NH)C6H4CO2CH3
CAS Number:
Molecular Weight:
165.19
Beilstein/REAXYS Number:
607217
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥95.0% (GC)

refractive index

n20/D 1.579 (lit.)
n20/D 1.580

bp

256 °C (lit.)

mp

17-19 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CNc1ccccc1C(=O)OC

InChI

1S/C9H11NO2/c1-10-8-6-4-3-5-7(8)9(11)12-2/h3-6,10H,1-2H3

InChI key

GVOWHGSUZUUUDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

216.7 °F - closed cup

flash_point_c

102.6 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hayato Nishida et al.
Regulatory toxicology and pharmacology : RTP, 72(3), 578-585 (2015-06-07)
Previously, a non-animal screening approach was proposed for evaluating photosafety of cosmetic ingredients by means of in vitro photochemical and photobiochemical assays; however, complex cosmetic ingredients, such as plant extracts and polymers, could not be evaluated because their molecular weight

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service