Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

657476

Sigma-Aldrich

(Z)-2-Bromo-2-butene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(Br)CH3
CAS Number:
Molecular Weight:
135.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.459 (lit.)

bp

91-92 °C (lit.)

density

1.331 g/mL at 25 °C (lit.)

Storage temp.

2-8°C

InChI

1S/C4H7Br/c1-3-4(2)5/h3H,1-2H3/b4-3-

Inchi Key

UILZQFGKPHAAOU-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

30.2 °F - closed cup

flash_point_c

-1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service