Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

657182

Sigma-Aldrich

2-Chloropyridium triflate on Wang resin

100-200 mesh particle size, extent of labeling: 1.0-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

Polymer-supported Mukaiyama reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.22

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carboxyl reactive

extent of labeling

1.0-1.5 mmol/g loading

particle size

100-200 mesh

Application

Used as an acid-activating agent and in the preparation of cabodiimides and guanidines.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and evaluation of a polymer-supported Mukaiyama reagent.
Emmanuelle Convers, Heather Tye and Mark Whittaker
Tetrahedron Letters, 45, 3401-3404 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service