About This Item
Recommended Products
assay
≥98.0% (GC)
≥98.0% (NT)
refractive index
n20/D 1.476
density
1.095 g/mL at 20 °C (lit.)
SMILES string
CNCC(O)CO
InChI
1S/C4H11NO2/c1-5-2-4(7)3-6/h4-7H,2-3H2,1H3
InChI key
WOMTYMDHLQTCHY-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
305.6 °F - closed cup
flash_point_c
152 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service