Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

656194

Sigma-Aldrich

Poly((9,9-dihexyl-9H-fluorene-2,7-vinylene)-co-(1-methoxy-4-(2-ethylhexyloxy)-2,5-phenylenevinylene)), 90:10 mole ratio

Synonym(s):

Poly(FV-co-MEHPV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C27H34)m(C17H24O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mp

>300 °C

solubility

chloroform: soluble
hexane: soluble

fluorescence

λex 281 nm; λem 499 nm in chloroform

Features and Benefits

Light emitting polymer with high efficiency and luminescence.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service