Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

656178

Sigma-Aldrich

3-(3-Methylphenyl)propionic acid

97%

Synonym(s):

3-Methylhydrocinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CC6H4CH2CH2CO2H
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

39-42 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1cccc(CCC(O)=O)c1

InChI

1S/C10H12O2/c1-8-3-2-4-9(7-8)5-6-10(11)12/h2-4,7H,5-6H2,1H3,(H,11,12)

Inchi Key

VWXVTHDQAOAENP-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service