Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655562

Sigma-Aldrich

Poly[1,6-bis(p-carboxyphenoxy)hexane]

Synonym(s):

Poly CPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H20O5)n
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

solubility

DMSO: soluble

General description

Poly[1,6-bis(p-carboxyphenoxy)hexane] (poly(CPH)) is a bioerodable polyanhydride.[1]

Application

Biodegradable polyanhydride with drug delivery applications.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of an injectable system based on bioerodible polyanhydride microspheres for sustained drug delivery.
Kipper M J, et al.
Biomaterials, 23(22), 4405-4412 (2002)
Neeraj Kumar et al.
Advanced drug delivery reviews, 54(7), 889-910 (2002-10-18)
Polyanhydrides have been considered to be useful biomaterials as carriers of drugs to various organs of the human body such as brain, bone, blood vessels, and eyes. They can be prepared easily from available, low cost resources and can be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service