Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654949

Sigma-Aldrich

Potassium 5-methyl-2-thiophenetrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C4H2SBF3K
CAS Number:
Molecular Weight:
204.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

95%

form

solid

mp

>220 °C
>250 °C (dec., DSC)

SMILES string

[K+].Cc1ccc(s1)[B-](F)(F)F

InChI

1S/C5H5BF3S.K/c1-4-2-3-5(10-4)6(7,8)9;/h2-3H,1H3;/q-1;+1

InChI key

SLHDDEQSKNIZEL-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Potassium trifluoroborates are moisture- and air-stable organoboron reagents suitable for oxidative conditions.

Related Content

The central theme of the Molander group's research is the development of new synthetic methods and their application to the synthesis of organic molecules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service