Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654264

Sigma-Aldrich

2-Bromopyridine-3-boronic acid pinacol ester

technical grade, 90%

Synonym(s):

2-Bromo-3-pyridineboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BBrNO2
CAS Number:
Molecular Weight:
283.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical grade

assay

90%

density

1.329 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2cccnc2Br

InChI

1S/C11H15BBrNO2/c1-10(2)11(3,4)16-12(15-10)8-6-5-7-14-9(8)13/h5-7H,1-4H3

InChI key

QAQDSIBZRCSYAR-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service