About This Item
assay
97%
form
solid
mp
80-84 °C (lit.)
SMILES string
Cc1cccc(n1)N2CCN(CC2)C(=O)OC(C)(C)C
InChI
1S/C15H23N3O2/c1-12-6-5-7-13(16-12)17-8-10-18(11-9-17)14(19)20-15(2,3)4/h5-7H,8-11H2,1-4H3
InChI key
QEDVZUFNZJJSJL-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service