Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65374

Sigma-Aldrich

4-Methoxytrityl chloride, polymer-bound

extent of labeling: ~0.7 mmol/g Cl loading

Synonym(s):

4-Methoxytriphenylchloromethane polymer-bound, MMT polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.7 mmol/g Cl loading

matrix

polystyrene, crosslinked with 1% DVB

particle size

200-400 mesh

application(s)

peptide synthesis

Looking for similar products? Visit Product Comparison Guide

Other Notes

Resin for solid-phase peptide synthesis and combinatorial chemistry. Cleavage occurs very mildly[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. van Vliet et al.
Proc. 22nd Eur. Pept. Symp.: Peptides 1992, 279-279 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service