Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

653438

Sigma-Aldrich

Poly(2,5-dioctyl-1,4-phenylenevinylene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C24H38)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

fluorescence

λex 406 nm; λem 490 nm in chloroform

General description

Poly(2,5-dioctyl-1,4-phenylenevinylene) (POPPV) is a poly(phenylenevinylene) derivative that is used as a green light emitting polymer. It is majorly utilized in the formation of organic electronic devices like polymer light emitting diodes (PLEDs).[1][2]

Physical properties

Light emitting conjugated polymer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoparticle electroluminescence: controlling emission color through Forster resonance energy transfer in hybrid particles
Huebner CF, et al.
Advances in Functional Materials, 19(22), 3604-3609 (2009)
Spectral tuning of conjugated polymer colloid light-emitting diodes
Huebner CF and Foulger SH
Langmuir, 26(4), 2945-2950 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service