About This Item
assay
98%
refractive index
n20/D 1.489 (lit.)
bp
47-49 °C/5 mmHg (lit.)
density
0.9113 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)c1ccccn1
InChI
1S/C8H13NSi/c1-10(2,3)8-6-4-5-7-9-8/h4-7H,1-3H3
InChI key
IBLKFJNZUSUTPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
125.6 °F - closed cup
flash_point_c
52 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service