Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

652474

Sigma-Aldrich

Carbon nanotube, single-walled

poly(ethylene glycol) functionalized, >80% carbonbasis, D × L 4-5 nm × 0.5-0.6 μm , bundle dimensions

Synonym(s):

Carbon nanotube, single-walled, PEG functionalized, SWNT, PEG functionalized

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

Carbon nanotube, single-walled, poly(ethylene glycol) functionalized, >80% carbon basis, D × L 4-5 nm × 0.5-0.6 μm , bundle dimensions, avg. no. of layers, 1

assay

>80% carbon basis

form

powder

mol wt

Mw ~600 g/mol (PEG)

feature

avg. no. of layers 1

D × L

4-5 nm × 0.5-0.6 μm , bundle dimensions

impurities

4-5% trace metals

solubility

H2O: 5 mg/mL

functional group

PEG

Preparation Note

Electric Arc Discharge Method
Procedure for dispersion of this PEG-functionalized SWNTs product in water:
Typically after this procedure, the material is completely dispersed according to visual observation.
Sonicate 50 mg of material in 5 mL water for 30 min. (A bath sonicator with a sonic power between 45 W and 270 W works fine. A sonic power of 270 W is preferable for faster dispersion).
Add 45 mL of water and sonicate for additional 60 min (or until the material appears well-dispersed).
Dilute the dispersion to the desired concentration and sonicate if necessary. If aggregates are still visible continue to sonicate until satisfactory dispersion is obtained. Note: If unable to achieve a satisfactory dispersion, reduce the initial amount of 50 mg of material in 5 mL of water.
This procedure is to be used as a basis and additional procedures can be researched. Results may vary if this is the first time working with this material. Solubility of this PEG-functionalized SWNTs product can reach 5.0 mg/mL in water.

Analysis Note

Carbonaceous content[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalized carbon nanomaterials: exploring the interactions with Caco-2 cells for potential oral drug delivery
Coyuco JC, et al.
International journal of nanomedicine, 6, 2253-2253 (2011)
Nano Letters, 3, 309-309 (2003)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

See All

Questions

  1. CNT의 나노파티클 크기가 몇 nm인지

    1 answer
    1. The dimensions of carbon nanotube bundles are found to be D×L 4-5nm × 0.5-0.6?m. They are described as 'nanotube bundles' rather than nanoparticles, which would have a diameter.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service