Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

652377

Sigma-Aldrich

1-Boc-4-(2-methoxycarbonylphenyl)piperazine

Synonym(s):

Methyl 2-(Boc-piperazin-1-yl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N2O4
CAS Number:
Molecular Weight:
320.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.5320 (lit.)

SMILES string

COC(=O)c1ccccc1N2CCN(CC2)C(=O)OC(C)(C)C

InChI

1S/C17H24N2O4/c1-17(2,3)23-16(21)19-11-9-18(10-12-19)14-8-6-5-7-13(14)15(20)22-4/h5-8H,9-12H2,1-4H3

InChI key

JFZGCXORLWXLGH-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of melanocortin-4 receptor antagonists

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service